Anti-CD223 (LAG-3) Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: C9B7W]

Supplier: Biolegend
125209 125210
125209-BLEA 105.9 USD
125209-BL 125210-BL
Anti-CD223 (LAG-3) Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: C9B7W]
Antibodies
APC anti-mouse CD223 (LAG-3) [C9B7W]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD223 (LAG-3)
Clonality: Monoclonal
Clone: C9B7W
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR