Anti-CD201 (EPCR) Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: RCR-401]

Supplier: Biolegend
351905 351906
351905-BLEA 127.91 USD
351905-BL 351906-BL
Anti-CD201 (EPCR) Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: RCR-401]
Antibodies
APC anti-human CD201 (EPCR) [RCR-401]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD201 (EPCR)
Clonality: Monoclonal
Clone: RCR-401
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR