Rtx®-440 Columns (Fused Silica), Restek
Supplier: Restek
12923 12924 12938 12939 42902
10850-436EA 815.38 USD
10850-436 10850-438 10850-440 10850-442 10850-444 10850-542
Rtx®-440 Columns (Fused Silica), Restek
Chromatography Columns
Rtx®-440 columns are useful general-purpose columns designed to be selective for pesticides, PAHs, and other semi-volatiles.
  • General-purpose columns with unique selectivity for pesticides, PAHs, or other semi-volatiles
  • Ideal for low/trace-level analyses
  • Low-bleed, high-resolution columns with unique selectivity
  • Wide temperature range: 20 to 340 °C
  • Mid-polarity proprietary Crossbond® phase
These fused silica columns are suitable for use over a wide temperature range, and are ideal for low-trace-level analyses.