Stx®-CLPesticides2 Columns (fused silica with Siltek® deactivation), Restek
Supplier: Restek
11444 11440 11441 11443 11444-600
RK11444EA 904.28 USD
RK11444 10850-014 10850-016 10850-018 10851-960
Stx®-CLPesticides2 Columns (fused silica with Siltek® deactivation), Restek
Chromatography Columns
Siltek® passivation technology enables these columns to offer unsurpassed inertness and the highest responses for active pesticides.
  • Application-specific columns for organochlorine pesticides and herbicides
  • Baseline separations in less than 10 minutes
  • Siltek® surface deactivation enhances responses for endrin, DDT, methoxychlor
  • Stable to 330 °C
  • Proprietary Crossbond® phases
Many laboratories analyzing organochlorine pesticides struggle with breakdown and adsorption of endrin, DDT, and methoxychlor caused by active sites throughout the analytical system, problems which are addressed by these columns.