Anti-CD123 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: 6H6]

Supplier: Biolegend
306019 306020
306019-BLEA 260.2 USD
306019-BL 10757-940
Anti-CD123 Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: 6H6]
Antibodies
Brilliant Violet 650™ anti-human CD123 [6H6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD123
Clonality: Monoclonal
Clone: 6H6
Conjugation: Brilliant Violet® 650
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR