Anti-CD10 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: HI10a]

Supplier: Biolegend
312215 312216
312215-BLEA 198.1 USD
312215-BL 312216-BL
Anti-CD10 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: HI10a]
Antibodies
PerCP/Cy5.5 anti-human CD10 [HI10a]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin monkey, Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: HI10a
Conjugation: PerCP (More Product Information
Order Now

SPECIFICATIONS

Antigen Symbol CD10
Environmentally Preferable
Antigen Synonyms Enkephalinase|Neutral endopeptidase|MME|Common acute lymphoblastic leukemia antigen (CALLA)|Neprilysin
Antibody Type Primary
Clonality Monoclonal
Conjugation PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Clone HI10a
Reactivity Baboon,Capuchin monkey,African green monkey,Cynomolgus monkey,Rhesus monkey,Chimpanzee,Human
Host Mouse
Gene ID 4311
Isotype IgG1 kappa
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated antibody.
Storage Temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping Temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR