BBL™ Isolator Cleanroom, eXtended Temperature (IC-XT) TSA Plates, Sterile Packs, BD Diagnostics
Supplier: BD
BBL™ IC-XT packs are gamma irradiated sterile plated media for environmental microbiology applications.
- Storage temperature range of 2 to 25 °C provides improved flexibility and resource utilization
- RODAC™ Snap Lid plates reduce risk during sampling and handling
- Shelf life of up to 6 months maximizes batch size and reduces incoming QC testing
- Robust sleeve material with desiccant reduces environmental effects on the media
- SAL claim of 10-6 minimizes risk of false positives and protects environment from risk of contaminants
- Triple-wrapped packaging enables easy transfer and storage while maintaining sterility
- Validated VHP resistance of the sleeve material and hanging holes on each bag improve handling in isolator
- Opening and removal from packaging is simple, even with isolator gloves
The BBL™ IC-XT plates are particularly useful for monitoring surfaces and air in Isolator Systems, clean rooms, and other environmentally controlled areas when medium sterility is important.