Merit™ Shamberg Comedone Extractor, Physician Grade, Sklar
Supplier: Sklar
98-740 98-741 98-742
10185-164EA 27.02 USD
10185-164 10185-166 75996-284
Merit™ Shamberg Comedone Extractor, Physician Grade, Sklar
Dissection Needles
The Merit™ Schamberg Comedone Extractor is a double-ended dermal instrument used to treat facial blemishes.
  • Fenestrated tips on both ends
  • Available in three sizes
  • Reusable and nonsterile
  • Latex-free
When placed around the blemish, gentle pressure is applied and the instrument assists in forcing the underlying debris out from beneath the skin.
Merit™ instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures.
The Schamberg Comedone Extractor is available in three varieties, tailoredMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable