Chloramphenicol 10 mg/ml, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol

J67273-AB J67273-AC
10814-950EA 91.3 USD
10814-950 10814-952
Chloramphenicol 10 mg/ml, sterile filtered
Chloramphenicol

Chloramphenicol, 10 mg/mL in ethanol, sterile-filtered.


Liquid


Precaución: Research use only: Not intended for animal or human diagnostic or therapeutic use.

Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00078159
CAS Number: 56-75-7
EINECS: 200-287-4
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR