D-Gulonic acid-γ-lactone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (3R,4S,5S)-5-((R)-1,2-Dihydroxyethyl)-3,4-dihydroxydihydrofuran-2(3H)-one, D-Gulono γ-lactone, (3R,4S,5S)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one, D-(-)-Gulono-1,4-lactone, D-Gulono-1,4-lactone

J66987-06 J66987-14
10816-032EA 57.52 USD
10816-032 10816-034
D-Gulonic acid-γ-lactone 98%
D-Gulonic acid-γ-lactone

Purity: 98% CAS No.: 6322-07-2 Molecular Formula: C6H10O6 Formula Weight: 178.14 Storage Temperature: 2°C to 8°C Physical Form: Powder Melting Point: 182-188° Solubility: Soluble in water Beilstein Reference: 83012 MDL No.: MFCD00005391

Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Melting Pt: 182…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005391
CAS Number: 6322-07-2
EINECS: 228-678-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR