1,2,3,4,6-Penta-O-acetyl-D-mannopyranose 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
alpha,beta-D-Mannose pentaacetate, 1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, (3S,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate
J66973-06
J66973-14
J66973-22
10815-954EA
34.4
USD
10815-954
10815-956
10815-958
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose 98%
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose
Purity: 98%
CAS No.: 25941-03-1
Molecular Formula: C16H22O11
Formula Weight: 390.34
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Melting Point: 66-72°
Optical Rotation: +54°(c=1 in chloroform)
MDL No.: MFCD05864874
Formula:
C₁₆H₂₂O₁₁ Melting Pt: 66…72 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05864874 CAS Number: 25941-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...