3,4-Di-O-acetyl-6-deoxy-L-glucal 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
3,4-Di-O-acetyl-1,5-anhydro-2,6-dideoxy-L-arabino-hex-1-enitol, 3,4-Di-O-acetyl-L-rhamnal
J66944-03
J66944-06
10815-916EA
48
USD
10815-916
10815-918
3,4-Di-O-acetyl-6-deoxy-L-glucal 98%
3,4-Di-O-acetyl-6-deoxy-L-glucal
Purity: 98%
CAS No.: 34819-86-8
Molecular Formula: C10H14O5
Formula Weight: 214.22
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₀H₁₄O₅ Boiling Pt: 68…69 °C Density: 1.116 Flash Pt: >110 °C (235 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00074970 CAS Number: 34819-86-8 EINECS: 252-228-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...