L-(+)-Gulonic acid-γ-lactone 95%

Supplier: Thermo Scientific Chemicals

Synonyms: L-Gulonic acid-1,4-lactone, (3S,4R,5R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxydihydrofuran-2(3H)-one, L-Gulonolactone

J66934-06 J66934-14
10815-906EA 50.7 USD
10815-906 10815-908
L-(+)-Gulonic acid-γ-lactone 95%
L-(+)-Gulonic acid-gamma-lactone

Purity: 95% CAS No.: 1128-23-0 Molecular Formula: C6H10O6 Formula Weight: 178.14 Storage Temperature: 2°C to 8°C Physical Form: Powder Melting Point: 184-190° Solubility: Soluble in water Beilstein Reference: 83002 Optical Rotation: +54.5° (c=4 in water) MDL No.: MFCD00064331

Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Melting Pt: 184…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064331
CAS Number: 1128-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR