5-Keto-D-gluconic acid potassium salt 98%

Supplier: Thermo Scientific Chemicals
Warning

J66916-03 J66916-06
10814-914EA 69.6 USD
10814-914 10814-916
5-Keto-D-gluconic acid potassium salt 98%
5-Keto-D-gluconic acid potassium salt

Purity: 98% CAS No.: 5447-60-9 Molecular Formula: C6H9KO7 Formula Weight: 232.23 Storage Temperature: 2°C to 8°C Physical Form: Powder Solubility: Soluble in water 50mg/ml Beilstein Reference: 3919618 MDL No.: MFCD00069562

Formula: C₆H₉KO₇
MW: 232.229 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00069562
CAS Number: 5447-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR