Tri-O-acetyl-D-galactal
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3R,4R)-2-(Acetoxymethyl)-3,4-dihydro-2H-pyran-3,4-diyl diacetate, 1,5-Anhydro-2-deoxy-D-lyxo-hex-1-enitol 3,4,6-Tri-O-acetyl Ether, 3,4,6-Tri-O-acetyl-D-galactal
J66904-03
J66904-06
J66904-09
10814-898EA
40.5
USD
10814-898
10814-900
10814-902
Tri-O-acetyl-D-galactal
Tri-O-acetyl-D-galactal
CAS No.: 4098-06-0
Molecular Formula: C12H16O7
Formula Weight: 272.25
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₂H₁₆O₇ Melting Pt: 34…38 °C Flash Pt: >110 °C(235 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00064092 CAS Number: 4098-06-0 EINECS: 223-859-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...