n-Dodecyl-β-D-maltoside

Supplier: Thermo Scientific Chemicals

Synonyms: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside

J66869-03 J66869-06 J66869-ME
10814-872EA 181.87 USD
10814-872 10814-874 10814-876
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside

A non-ionic detergent useful in membrane solubilization studies.


CAS No.: 69227-93-6 Molecular Formula: C24H46O11 Formula Weight: 510.62 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Melting Point: 224-226° Solubility: Soluble in water at 10% w/v Beilstein Reference: 55318 MDL No.: MFCD00043012


A non-ionic detergent useful in membrane solubilization studies

Formula: C₂₄H₄₆O₁₁
MW: 510.6 g/mol
Melting Pt: 224…226 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043012
CAS Number: 69227-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR