2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
Supplier: Thermo Scientific Chemicals
J66853-06
J66853-09
10814-862EA
45.09
USD
10814-862
10814-864
10814-866
2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
CAS No.: 60933-68-8
Molecular Formula: C26H28O5
Formula Weight: 420.50
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in methanol
MDL No.: MFCD00047076
Formula:
C₂₆H₂₈O₅ MW: 420.505 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00047076 CAS Number: 60933-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...