Sucrose stearate

Supplier: Thermo Scientific Chemicals

J66845-06 J66845-14
10815-894EA 42.6 USD
10815-894 10815-896
Sucrose stearate
Sucrose stearate

CAS No.: 25168-73-4 Molecular Formula: C30H56O12 Formula Weight: 608.75 Physical Form: Powder Appearance: White to off-white

Formula: C₃₀H₅₆O₁₂
MW: 608.766 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152822
CAS Number: 25168-73-4
EINECS: 246-705-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR