Methyl-β-D-ribofuranoside 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-methoxytetrahydrofuran-3,4-diol, Methyl β-D-Ribofuranoside
J66828-06
J66828-14
10815-876EA
67.6
USD
10815-876
10815-878
Methyl-β-D-ribofuranoside 98%
Methyl-beta-D-ribofuranoside
Purity: 98%
CAS No.: 7473-45-2
Molecular Formula: C6H12O5
Formula Weight: 164.16
Physical Form: Powder
Appearance: White to off-white
Melting Point: 78°
Solubility: Soluble in water
Optical Rotation: -50° (c=2 in water)
MDL No.: MFCD00047075
Formula:
C₆H₁₂O₅ MW: 164.16 g/mol Melting Pt: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047075 CAS Number: 7473-45-2 EINECS: 231-271-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...