(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-α-D-arabinofuranose, 2-Deoxy-2-fluoro-1,3,5-tri-O-benzoyl-α-D-arabinofuranose

J66805-03 J66805-06
10815-840EA 50.4 USD
10815-840 10815-842
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate

CAS No.: 97614-43-2 Molecular Formula: C26H21FO7 Formula Weight: 464.44 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Melting Point: 76-78° Solubility: Soluble in chloroform or dichloromethane MDL No.: MFCD00083339

Formula: C26H21FO7
MW: 464.44 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00083339
CAS Number: 97614-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR