Cecropin A (2-9) Peptide

Supplier: Thermo Scientific Chemicals

J66741-EXD J66741-MCR
10814-820EA 149 USD
10814-820 10814-822
Cecropin A (2-9) Peptide
Proteins and Peptides

Molecular Formula: C89H152N22O15
Formula Weight: 1770.29
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
Solubility: Soluble in water at 1mg/ml
MDL No.: MFCD01630779


Causes the release of carboxyfluoresceine encapsulated in phosphatidylcholine liposomes.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR