Lactose octaacetate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3R,4S,5R,6R)-6-(Acetoxymethyl)-5-(((2S,3R,4S,5S,6R)-3,4,5-triacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2,3,4-triyl triacetate

J66695-06 J66695-14
10814-774EA 38.9 USD
10814-774 10814-776
Lactose octaacetate 98%
Lactose octaacetate

Purity: 98% CAS No.: 6291-42-5 Molecular Formula: C28H38O19 Formula Weight: 678.59 Storage Temperature: -30°C to -10°C Physical Form: Powder Solubility: Soluble in chloroform MDL No.: MFCD00064040

Formula: C₂₈H₃₈O₁₉
MW: 678.593 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00064040
CAS Number: 6291-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR