1,2,3,4,6-Penta-O-acetyl-α-D-mannopyranose 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3S,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate, a-D-Mannose pentaacetate
J66685-06
J66685-14
10814-758EA
82.6
USD
10814-758
10814-760
1,2,3,4,6-Penta-O-acetyl-α-D-mannopyranose 98%
1,2,3,4,6-Penta-O-acetyl-alpha-D-mannopyranose
Purity: 98%
CAS No.: 4163-65-9
Molecular Formula: C16H22O11
Formula Weight: 390.34
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Solubility: Soluble in chloroform
Optical Rotation: +54° (c=1 in chloroform)
MDL No.: MFCD00069798
Formula:
C₁₆H₂₂O₁₁ Storage Temperature: Freezer |
MDL Number:
MFCD00069798 CAS Number: 4163-65-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...