D-Galacto-D-mannan
Supplier: Thermo Scientific Chemicals
Synonyms:
D-Galacto-D-mannane, Galactomannoglycan, Carobgalactomannan
J66610-MC
J66610-ME
10815-718EA
116
USD
10815-718
10815-720
D-Galacto-D-mannan
D-Galacto-D-mannan
CAS No.: 11078-30-1
Formula Weight: ~200000
Physical Form: Powder
Appearance: White
MDL No.: MFCD00146683
Formula:
C₁₈H₃₂O₁₆ MW: 504.437 Da Storage Temperature: Ambient |
MDL Number:
MFCD00146683 CAS Number: 11078-30-1 EINECS: 234-299-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...