2-O-ɑ-D-glucopyranosyl-L-ascorbic acid 99.5%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-5-((S)-1,2-Dihydroxyethyl)-4-hydroxy-3-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)furan-2(5H)-one, AA-2G, Ascorbic acid 2-glucoside
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduces the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. 2-O-alpha-D-Glucopyranosyl-L-ascorbic acid also promotes collagen synthesis and function as an antioxidant.
Purity: 99.5%
CAS No.: 129499-78-1
Molecular Formula: C12H18O11
Formula Weight: 338.26
Physical Form: Powder
Appearance: White to off-white
Melting Point: 158-163°
MDL No.: MFCD23701380
Formula:
C₁₂H₁₈O₁₁ MW: 338.27 g/mol Melting Pt: 158…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD23701380 CAS Number: 129499-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...