2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
D-Fructopyranose diacetonide, ((3aS,5aR,8aR,8bS)-2,2,7,7-Tetramethyltetrahydro-3aH-bis([1,3]dioxolo)[4,5-b:4',5'-d]pyran-3a-yl)methanol, Diacetonefructose
J66537-09
J66537-18
J66537-22
10814-686EA
46.3
USD
10814-686
10814-688
10814-690
2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose 98%
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose
Purity: 98%
CAS No.: 20880-92-6
Molecular Formula: C12H20O6
Formula Weight: 260.28
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Melting Point: 93-96°
Optical Rotation: -25.5°(c=1 in chloroform)
MDL No.: MFCD00022183
Formula:
C₁₂H₂₀O₆ MW: 260.29 g/mol Melting Pt: 93…96 °C Storage Temperature: Freezer |
MDL Number:
MFCD00022183 CAS Number: 20880-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...