maltotriose 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal
Maltotriose is a matrose regulon inducer in Escherichia coli.
Maltotriose has been used in a study to investigate the influence of fourth generation poly(propyleneimine) dendrimers on blood cells.
Precaución: Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Formula:
C₁₈H₃₂O₁₆ MW: 504.438 g/mol Melting Pt: 132…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006629 CAS Number: 1109-28-0 EINECS: 214-174-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...