1-Deoxynojirimycin hydrochloride 98%
Supplier: Thermo Scientific Chemicals
J66458-MA
J66458-MB
AAJ66458-MAEA
154
USD
AAJ66458-MA
10814-658
1-Deoxynojirimycin hydrochloride 98%
1-Deoxynojirimycin hydrochloride
White powder
Soluble in water at 20 mg/mL
Formula:
C₆H₁₃NO₄·HCl Storage Temperature: Refrigerator |
MDL Number:
MFCD00133247 CAS Number: 73285-50-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...