2,3,6,2',3',4',6'-Hepta-O-acetyl-α-D-lactosyl bromide
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2R,3S,4S,5R,6S)-2-(Acetoxymethyl)-6-(((2R,3R,4S,5R,6R)-4,5-diacetoxy-2-(acetoxymethyl)-6-bromotetrahydro-2H-pyran-3-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate, 2,2',3,3',4',6,6'-HEPTA-O-ACETYL-α-D-LACTOSYL BROMIDE, 2,3,6,2 ,3 ,4 ,6 -Hepta-O-acetyl-a-D-cellobiosyl bromide, Bromo Heptaacetyl-D-lactoside, Acetobromo-α-D-lactoside
CAS No.: 4753-07-5
Molecular Formula: C26H35BrO17
Formula Weight: 699.45
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to off-white
Solubility: Soluble in chloroform
MDL No.: MFCD00153593
Formula:
C₂₆H₃₅BrO₁₇ MW: 699.45 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00153593 CAS Number: 4753-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...