α-D-Galactose pentaacetate
Supplier: Thermo Scientific Chemicals
J66366-06
J66366-14
10814-578EA
33.1
USD
10814-578
10814-580
10814-582
α-D-Galactose pentaacetate
alpha-D-Galactose pentaacetate
CAS No.: 4163-59-1
Molecular Formula: C16H22O11
Formula Weight: 390.34
Physical Form: Powder
Appearance: White to off-white
Melting Point: 143-144°
MDL No.: MFCD00064081
Formula:
C₁₆H₂₂O₁₁ Melting Pt: 143…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064081 CAS Number: 4163-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...