2,3-O-Isopropylidene-D-glyceraldehyde 50% (w/w) in dichloromethane

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde, (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde, (R)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde

J66337-03 J66337-06 J66337-09
10815-572EA 79.8 USD
10815-572 10815-574 10815-576
2,3-O-Isopropylidene-D-glyceraldehyde 50% (w/w) in dichloromethane
2,3-O-Isopropylidene-D-glyceraldehyde

CAS No.: 15186-48-8 Molecular Formula: C6H10O3 Formula Weight: 130.14 Storage Temperature: 2°C to 8°C Flash Point: 62°(144°F) Physical Form: Liquid Optical Rotation: +53.8° (c=2 in chloroform) UN No.: 1593 MDL No.: MFCD00269682

Formula: C₆H₁₀O₃
MW: 130.143 g/mol
Flash Pt: 62 °C (144 °F)
Storage Temperature: Ambient
MDL Number: MFCD00269682
CAS Number: 15186-48-8
UN: 1593
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR