D-Fructose-6-phosphate disodium salt 95%

Supplier: Thermo Scientific Chemicals
Warning

J66311-03 J66311-MC J66311-ME
10815-538EA 150 USD
10815-538 10815-540 10815-542
D-Fructose-6-phosphate disodium salt 95%
D-Fructose-6-phosphate disodium salt

Purity: 95% CAS No.: 26177-86-6 Molecular Formula: C6H11Na2O9P Formula Weight: 304.10 Storage Temperature: -30°C to -10°C Physical Form: Powder MDL No.: MFCD00135619

Formula: C₆H₁₁Na₂O₉P
MW: 304.098 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00135619
CAS Number: 26177-86-6
EINECS: 247-505-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR