1,2,3,4-Tetra-O-acetyl-β-D-glucopyranose 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Hexopyranose, 1,2,3,4-tetraacetate, 1,2,3,4-Tetra-o-acetyl-beta-d-glucopyranose
J66307-03
J66307-ME
10815-534EA
160
USD
10815-534
10815-536
1,2,3,4-Tetra-O-acetyl-β-D-glucopyranose 98%
1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose
Purity: 98%
CAS No.: 13100-46-4
Molecular Formula: C14H20O10
Formula Weight: 348.30
Physical Form: Powder
Appearance: White to off-white
Melting Point: 126-128°
Solubility: Soluble in chloroform
Optical Rotation: +8° (c=6 in chloroform)
MDL No.: MFCD00063260
Formula:
C₁₄H₂₀O₁₀ MW: 348.304 g/mol Melting Pt: 126…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063260 CAS Number: 13100-46-4 EINECS: 236-018-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...