Methyl-2,3-O-isopropylidene-β-D-ribofuranoside 98%

Supplier: Thermo Scientific Chemicals

Synonyms: ((3aR,4R,6R,6aR)-6-Methoxy-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)methanol

J66296-03 J66296-06
10815-498EA 52.3 USD
10815-498 10815-500
Methyl-2,3-O-isopropylidene-β-D-ribofuranoside 98%
Methyl-2,3-O-isopropylidene-beta-D-ribofuranoside

Purity: 98% CAS No.: 4099-85-8 Molecular Formula: C9H16O5 Formula Weight: 204.22 Storage Temperature: 2°C to 8°C Physical Form: Oil Solubility: Soluble in chloroform, dichloromethane, ethanol, ethyl acetate, and methanol Optical Rotation: -77° (c=1 in dichloromethane) MDL No.: MFCD00270040

Formula: C₉H₁₆O₅
MW: 204.222 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270040
CAS Number: 4099-85-8
EINECS: 223-865-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR