α-D-Galacturonic acid monohydrate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2S,3R,4S,5R)-2,3,4,5-Tetrahydroxy-6-oxohexanoic acid hydrate, D-Galacturonic acid monohydrate
J66282-06
J66282-14
10815-486EA
49.75
USD
10815-486
10815-488
α-D-Galacturonic acid monohydrate 97%
alpha-D-Galacturonic acid monohydrate
D-(+)-Galacturonic acid monohydrate can be used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.
Purity: 97%
CAS No.: 91510-62-2
Molecular Formula: C6H10O7.H2O
Formula Weight: 212.15
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water
Beilstein Reference: 1727087
Merck Reference: 14,4337
Optical Rotation: +53° (c=10 in water)
MDL No.: MFCD00071585
Formula:
C₆H₁₀O₇.H₂O MW: 212.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00071585 CAS Number: 91510-62-2 EINECS: 211-682-6 Merck Index: 14,4337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...