Calcium-L-threonate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Calcium L-Threonate, L-Threonic acid Calcium salt, Calcium (2R,3S)-2,3,4-trihydroxybutanoate
J66279-22
J66279-36
10815-478EA
81.1
USD
10815-478
10815-480
10815-482
Calcium-L-threonate 98%
Calcium-L-threonate
Purity: 98%
CAS No.: 70753-61-6
Molecular Formula: C8H14CaO10
Formula Weight: 310.27
Physical Form: Powder
Appearance: Pale brown
Melting Point:
Formula:
C₄H₈O₅ MW: 310.27 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150824 CAS Number: 70753-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...