D-Glucosamine sulfate 98%
Supplier: Thermo Scientific Chemicals
J66271-06
J66271-14
10814-554EA
46.4
USD
10814-554
10815-476
D-Glucosamine sulfate 98%
D-Glucosamine sulfate
Purity: 98%
CAS No.: 29031-19-4
Molecular Formula: C6H13NO5.H2SO4
Formula Weight: 277.25
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble at 1mg/ml in 1M HCl
MDL No.: MFCD00080790
Formula:
C₆H₁₅NO₉S MW: 277.244 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00080790 CAS Number: 29031-19-4 EINECS: 249-379-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...