D(+)-Ribonic acid γ-lactone
Supplier: Thermo Scientific Chemicals
Synonyms:
(3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one, Ribono-γ-lactone, D-Ribonic acid-1,4-lactone
J66266-03
J66266-06
10814-550EA
56.2
USD
10814-550
10814-552
D(+)-Ribonic acid γ-lactone
D(+)-Ribonic acid γ-lactone
CAS No.: 5336-08-3
Molecular Formula: C5H8O5
Formula Weight: 148.11
Physical Form: Powder
Appearance: White to pale yellow
Melting Point: 85-87°
Beilstein Reference: 82057
MDL No.: MFCD00063241
Formula:
C₅H₈O₅ MW: 148.11 g/mol Melting Pt: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063241 CAS Number: 5336-08-3 EINECS: 226-256-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...