Acrylamide 99.5% for molecular biology

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Prop-2-enamide

J66184-A1 J66184-36
10815-470EA 336.83 USD
10815-470 10814-486
Acrylamide 99.5% for molecular biology
Acrylamide (monomer)

A highly purified acrylamide for superior gel formation. Useful in sensitive small molecule applications

Formula: CH₂=CHCONH₂
MW: 71.08 g/mol
Boiling Pt: 125 °C/25 mm
Melting Pt: 82…85 °C
Density: 1.322
Flash Pt: 138 °C(280 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: "6.1",III
Merck Index: 14,129
https://pr.vwr.com/stibo/web/std.lang.all/13/21/2301321.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR