4-Methylumbelliferyl sulfate potassium salt 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Potassium 4-methyl-2-oxo-2H-chromen-7-yl sulphate, 4-Methylumbelliferyl sulphate potassium
J66160-03
J66160-ME
10814-460EA
231
USD
10814-460
10814-462
10814-464
4-Methylumbelliferyl sulfate potassium salt 98%
4-Methylumbelliferyl sulfate potassium salt
Substrate for sulfatase
Purity: 98%
CAS No.: 15220-11-8
Molecular Formula: C10H7KO6S
Formula Weight: 294.32
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Solubility: Soluble in water at 5mg/ml
Beilstein Reference: 3803203
MDL No.: MFCD00016970
Substrate for sulfatase
Formula:
C₁₀H₇KO₆S MW: 294.318 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00016970 CAS Number: 15220-11-8 EINECS: 239-272-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...