Bovine [Glp1]-Apelin-13

Supplier: Thermo Scientific Chemicals

J66114-EXD J66114-MCR
10815-454EA 85.4 USD
10815-454 10815-456
Bovine [Glp1]-Apelin-13
Proteins and Peptides

Molecular Formula: C69H108N22O16S
Formula Weight: 1533.79
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR