1,2-O-Isopropylidene-ɑ-D-xylofuranose 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1,2-O-Isopropylidene-α-D-xylofuranose, (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol, (3aR,5R,6S,6aR)-5-(Hydroxymethyl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol
J66110-06
J66110-09
J66110-14
10815-448EA
70.8
USD
10815-448
10815-450
10815-452
1,2-O-Isopropylidene-ɑ-D-xylofuranose 98%
1,2-O-Isopropylidene-alpha-D-xylofuranose
Purity: 98%
CAS No.: 20031-21-4
Molecular Formula: C8H14O5
Formula Weight: 190.19
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Melting Point: 69-71°
Boiling Point: 112-114°/0.06mm
Beilstein Reference: 81520
Optical Rotation: -19°(c=1 in water)
MDL No.: MFCD00063295
Formula:
C₈H₁₄O₅ MW: 190.20 g/mol Boiling Pt: 112…114 °C/0.06 mm Melting Pt: 69…71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063295 CAS Number: 20031-21-4 EINECS: 606-426-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...