N-Acetyl-D-galactosamine
Supplier: Thermo Scientific Chemicals
CAS No.: 1811-31-0
Molecular Formula: C8H15NO6
Formula Weight: 221.21
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
Melting Point: 160°
Solubility: Soluble in water at 50mg/ml
Beilstein Reference: 2331340
MDL No.: MFCD00065372
Formula:
C₈H₁₅NO₆ Melting Pt: 160 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00065372 CAS Number: 1811-31-0 EINECS: 217-321-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...