2,3,4,6-Tetra-O-acetyl-D-glucopyranose 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
2,3,4,6-tetra-O-acetylglucopyranose, D-Glucopyranose 2,3,4,6-tetraacetate, (2R,3R,4S,5R)-2-(Acetoxymethyl)-6-hydroxytetrahydro-2H-pyran-3,4,5-triyl triacetate, 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranose
J66052-03
J66052-06
J66052-09
10815-424EA
61.7
USD
10815-424
10815-426
10815-428
2,3,4,6-Tetra-O-acetyl-D-glucopyranose 98%
2,3,4,6-Tetra-O-acetyl-D-glucopyranose
Purity: 98%
CAS No.: 10343-06-3
Molecular Formula: C14H20O10
Formula Weight: 348.30
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Melting Point: 137-138°
Solubility: Soluble in chloroform or ethyl acetate
MDL No.: MFCD00061634
Formula:
C₁₄H₂₀O₁₀ Melting Pt: 137…138 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00061634 CAS Number: 10343-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...