4-Nitrophenyl-ɑ-D-mannopyranoside 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Nitrophenyl α-D-Mannopyranoside, 4-Nitrophenyl a-D-mannopyranoside
J66026-03
J66026-ME
10815-398EA
185
USD
10815-398
10815-400
4-Nitrophenyl-ɑ-D-mannopyranoside 98%
4-Nitrophenyl-alpha-D-mannopyranoside
Purity: 98%
CAS No.: 10357-27-4
Molecular Formula: C12H15NO8
Formula Weight: 301.25
Physical Form: Powder
Appearance: White to off-white
Melting Point: 178-183°
Beilstein Reference: 92210
MDL No.: MFCD00066002
Formula:
C₁₂H₁₅NO₈ MW: 301.25 g/mol Melting Pt: 178…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066002 CAS Number: 10357-27-4 EINECS: 233-776-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...