2,3,6,2',3',4',6'-Hepta-O-acetyl-α-D-cellobiosyl bromide
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-(((2R,3R,4S,5R,6R)-4,5-diacetoxy-2-(acetoxymethyl)-6-bromotetrahydro-2H-pyran-3-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate, Acetobromocellobiose
J66024-06
J66024-09
10815-388EA
29.49
USD
10815-388
10815-390
10815-392
2,3,6,2',3',4',6'-Hepta-O-acetyl-α-D-cellobiosyl bromide
2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide
CAS No.: 14227-66-8
Molecular Formula: C26H35BrO17
Formula Weight: 699.45
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to off-white
Melting Point: 180-190°
Solubility: Soluble in water. Slightly soluble in ethanol
MDL No.: MFCD00069839
Formula:
C₂₆H₃₅BrO₁₇ MW: 699.453 g/mol Melting Pt: 180…190 °C Storage Temperature: Freezer |
MDL Number:
MFCD00069839 CAS Number: 14227-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...