D-Glucose-6-phosphate disodium salt 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
D-Glucose 6-phosphate disodium salt, Sodium (2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl phosphate, Robison Ester Disodium Salt
J66023-03
J66023-06
10815-384EA
72.62
USD
10815-384
10815-386
D-Glucose-6-phosphate disodium salt 98%
D-Glucose-6-phosphate disodium salt
Purity: 98%
CAS No.: 3671-99-6
Molecular Formula: C6H11Na2O9P
Formula Weight: 304.10
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Solubility: Soluble in water
MDL No.: MFCD00136037
Formula:
C₆H₁₁Na₂O₉P Storage Temperature: Refrigerator |
MDL Number:
MFCD00136037 CAS Number: 3671-99-6 EINECS: 222-938-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...