D-Ribose-5-phosphate barium salt hexahydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

J66017-03 J66017-MD
10815-376EA 339 USD
10815-376 10815-378
D-Ribose-5-phosphate barium salt hexahydrate 99%
D-Ribose-5-phosphate barium salt hexahydrate

Purity: 99% CAS No.: 15673-79-7 Molecular Formula: C5H9BaO8P.6H2O Formula Weight: 473.51 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to pale yellow Beilstein Reference: 3781061 Optical Rotation: +12° (c=1 in 0.2M HCl) MDL No.: MFCD00149441

Formula: C₅H₁₁O₈P·6H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00149441
CAS Number: 15673-79-7
EINECS: 239-750-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR