Nonyl-β-D-glucopyranoside 98%
Supplier: Thermo Scientific Chemicals
J66009-03
J66009-06
10815-364EA
260
USD
10815-364
10815-366
Nonyl-β-D-glucopyranoside 98%
Nonyl-β-D-glucopyranoside
Purity: 98%
CAS No.: 69984-73-2
Molecular Formula: C15H30O6
Formula Weight: 306.40
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Beilstein Reference: 12168
MDL No.: MFCD00063300
Formula:
C₁₅H₃₀O₆ Storage Temperature: Freezer |
MDL Number:
MFCD00063300 CAS Number: 69984-73-2 EINECS: 615-042-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...