D-(+)-Trehalose, anhydrous, powder

Supplier: Thermo Scientific Chemicals

Synonyms: (+)-Trehalose, α,α-Trehalose, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol), Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside, D-(+)-Trehalose Anhydrous, D-Trehalose

J66006-06 J66006-09 J66006-18
10815-358EA 43.93 USD
10815-358 10815-360 10815-362
D-(+)-Trehalose, anhydrous, powder
D-(+)-Trehalose

CAS No.: 99-20-7 Molecular Formula: C12H22O11 Formula Weight: 342.30 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to off-white Optical Rotation: +197° (c=7 in water) MDL No.: MFCD00006628

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006628
CAS Number: 99-20-7
EINECS: 202-739-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR